Anti-TARDBP Mouse Monoclonal Antibody [clone: 2E2-D3]

Proveedor: US Biological
134223
USBI134223EA 655 EUR
USBI134223
Anti-TARDBP Mouse Monoclonal Antibody [clone: 2E2-D3]
Anticuerpos
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: TARDBP (TAR DNA binding protein)
Clonality: monoclonal
Clone: 2E2-D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno TARDBP
Environmentally Preferable
Nombre del antígeno TAR DNA binding protein
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 2E2-D3
Reactividad Human
Huésped Mouse
Isotipo IgG1 kappa
Western blot Yes
ELISA Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Sequence MSEYIRVTEDENDEPIEIPSEDDGTVLLSTVTAQFPGACGLRYRNPVSQCMRGVRLVEGILHAPDAGWGNLVYVVNYPKDNKRKMDETDASSAVKVKRAVQKTSDLIVLGLPWKTTEQDLKEYFSTFGEVLMVQVKKDLKTGHSKGFGFVRFTEYETQVKVMSQRHMIDGRWCDCKLPNSKQSQDEPLRSRKVFVGRCTEDMTEDELREFFSQYGDVMDVFIPKPFRAFAFVTFADDQIAQSLCGEDLIIKGISV
Forma liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR