Anti-PBP2a Mouse Monoclonal Antibody [clone: M8121521]

Proveedor: US Biological
138208
USBI138208EA 601 EUR
USBI138208
Anti-PBP2a Mouse Monoclonal Antibody [clone: M8121521]
Anticuerpos
Type: Primary
Antigen: PBP2a
Clonality: monoclonal
Clone: M8121521
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR