Anti-CLU Mouse Monoclonal Antibody [clone: 13j22]

Proveedor: US Biological
139770
USBI139770EA 543 EUR
USBI139770
Anti-CLU Mouse Monoclonal Antibody [clone: 13j22]
Anticuerpos
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: CLU (clusterin)
Clonality: monoclonal
Clone: 13j22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR