Anti-AGT Mouse Monoclonal Antibody [clone: 3C10]

Proveedor: US Biological
A2294-48Z
USBIA2294-48ZEA 624 EUR
USBIA2294-48Z
Anti-AGT Mouse Monoclonal Antibody [clone: 3C10]
Anticuerpos
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: AGT (angiotensinogen (serpin peptidase inhibitor, clade A, member 8))
Clonality: monoclonal
Clone: 3C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno AGT
Environmentally Preferable
Nombre del antígeno Angiotensinogen
Sinónimos de antígenos Member 8|Clade A|Serpin Peptidase Inhibitor|SERPINA8
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 3C10
Reactividad Human
Huésped Mouse
Isotipo IgG3 kappa
Western blot Yes
ELISA Yes
Número de aminoácidos 1 to 486
Forma liquid
Purification purified by Protein A chromatography
Concentración ~0,5 mg/ml
Storage buffer PBS, pH 7,2.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR