Anti-ANTXR1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Proveedor: US Biological
A2296-97C-AP
USBIA2296-97C-APEA 884 EUR
USBIA2296-97C-AP
Anti-ANTXR1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: ANTXR1 (anthrax toxin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR