Anti-BAD Rabbit polyclonal antibody unconjugated

Proveedor: US Biological
B0003-47M
USBIB0003-47MEA 667 EUR
USBIB0003-47M
Anti-BAD Rabbit polyclonal antibody unconjugated
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: BAD (BCL2-associated agonist of cell death)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno BAD
Nombre del antígeno BCL2-associated agonist of cell death
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
Modificación phospho-Ser134
Forma liquid
Purification purified by immunoaffinity chromatography
Storage buffer PBS, 0,09% sodium azide.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR