Anti-CA9 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: US Biological
C1105-80D-FITC
USBIC1105-80D-FITCEA 884 EUR
USBIC1105-80D-FITC
Anti-CA9 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno CA9
Nombre del antígeno carbonic anhydrase IX
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reactividad Human
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
Epítopo N-terminal
Forma liquid
Purification purified by Protein G chromatography
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Temperatura de almacenamiento Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR