Anti-Cholecystokinin 8 Rabbit Polyclonal Antibody

Proveedor: US Biological
C5042-10D
USBIC5042-10DEA 587 EUR
USBIC5042-10D
Anti-Cholecystokinin 8 Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: Immunochemistry: 1:200-1:5000

Type: Primary
Antigen: Cholecystokinin 8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno CCK8
Nombre del antígeno Cholecystokinin 8
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human,Rat,Mouse
Huésped Rabbit
Isotipo IgG
Formato Serum
Forma liquid
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR