Anti-CLK2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Proveedor: US Biological
C5841-59-PE
USBIC5841-59-PEEA 884 EUR
USBIC5841-59-PE
Anti-CLK2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: CLK2 (CDC-like kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR