Anti-COL6 Goat Polyclonal Antibody

Proveedor: US Biological
C7510-60M
USBIC7510-60MEA 681 EUR
USBIC7510-60M
Anti-COL6 Goat Polyclonal Antibody
Anticuerpos
Recommended dilution: Immunochemistry: 1:10-1:20;

Type: Primary
Antigen: COL6 (collagen, type VI)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR