Anti-GJA5 Mouse Monoclonal Antibody [clone: 6A47]

Proveedor: US Biological
C7856-03B
USBIC7856-03BEA 775 EUR
USBIC7856-03B
Anti-GJA5 Mouse Monoclonal Antibody [clone: 6A47]
Anticuerpos
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 1-3?g/ml; ELISA: 0.1-1 ?g/ml

Type: Primary
Antigen: GJA5 (gap junction protein, alpha 5, 40kDa)
Clonality: Monoclonal
Clone: 6A47
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR