Anti-CROT Rabbit polyclonal antibody

Proveedor: US Biological
C7904-43H
USBIC7904-43HEA 649 EUR
USBIC7904-43H
Anti-CROT Rabbit polyclonal antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CROT (carnitine O-octanoyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR