Anti-Factor H Mouse Monoclonal Antibody [clone: OX-24]

Proveedor: US Biological
F0010-01P
USBIF0010-01PEA 632 EUR
USBIF0010-01P
Anti-Factor H Mouse Monoclonal Antibody [clone: OX-24]
Anticuerpos
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Factor H
Clonality: monoclonal
Clone: OX-24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR