Anti-Fibrinogen Sheep Polyclonal Antibody (FITC (Fluorescein))

Proveedor: US Biological
F4203-02F1
USBIF4203-02F1EA 607 EUR
USBIF4203-02F1
Anti-Fibrinogen Sheep Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Recommended dilution: Immunofluorescence: 1:10-1:100

Type: Primary
Antigen: Fibrinogen (fibrinogen)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Nombre del antígeno Fibrinogen
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reactividad Human,Rat,Mouse
Huésped Sheep
Isotipo IgG
FluorescenciaInmunológica Yes
Forma liquid
Purification purified by immunoaffinity chromatography
Concentración ~1 mg/ml
Storage buffer PBS, pH 7,4, 0,09% sodium azide
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR