Anti-FOXO4 Rabbit Polyclonal Antibody

Proveedor: US Biological
F9049-45D
USBIF9049-45DEA 632 EUR
USBIF9049-45D
Anti-FOXO4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: FOXO4 (forkhead box O4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR