Anti-G Protein Coupled Bile Acid Receptor 1 Mouse monoclonal antibody [clone: 7H236]

Proveedor: US Biological
G8600-03H
USBIG8600-03HEA 817 EUR
USBIG8600-03H
Anti-G Protein Coupled Bile Acid Receptor 1 Mouse monoclonal antibody [clone: 7H236]
Anticuerpos
Type: Primary
Antigen: G Protein Coupled Bile Acid Receptor 1
Clonality: monoclonal
Clone: 7H236
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR