Anti-IgG Donkey Polyclonal Antibody (Rhodamine)

Proveedor: US Biological
I1904-45N7
USBII1904-45N7EA 243 EUR
USBII1904-45N7
Anti-IgG Donkey Polyclonal Antibody (Rhodamine)
Anticuerpos
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR