Anti-IgG/IgM Goat Polyclonal Antibody (Rhodamine)

Proveedor: US Biological
I1904-48F7
USBII1904-48F7EA 391 EUR
USBII1904-48F7
Anti-IgG/IgM Goat Polyclonal Antibody (Rhodamine)
Anticuerpos
Type: Primary
Antigen: IgG/IgM (IgG/IgM)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR