Anti-IKBKB Mouse Monoclonal Antibody [clone: 8L373]

Proveedor: US Biological
I3000-08J
USBII3000-08JEA 613 EUR
USBII3000-08J
Anti-IKBKB Mouse Monoclonal Antibody [clone: 8L373]
Anticuerpos
Recommended dilution: Western Blot: 5 ?g/ml

Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: monoclonal
Clone: 8L373
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno IKBKB
Nombre del antígeno Inhibitor of Kappa Light Polypeptide Gene Enhancer in B-cells, kinase beta
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 8L373
Reactividad Human,Mouse
Huésped Mouse
Isotipo IgG1 kappa
Western blot Yes
Precipitación inmunológica Yes
Forma liquid
Purification purified by Protein G chromatography
Concentración ~0,5 mg/ml
Storage buffer PBS, 0,2% gelatin, 0,05% sodium azide.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR