Anti-IKK alpha, beta Rabbit Polyclonal Antibody

Proveedor: US Biological
I3000-11A
USBII3000-11AEA 379 EUR
USBII3000-11A
Anti-IKK alpha, beta Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:4000; Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: IKK alpha, beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR