Anti-Influenza B Mouse Monoclonal Antibody [clone: 8J123]

Proveedor: US Biological
I7651-16D
USBII7651-16DEA 512 EUR
USBII7651-16D
Anti-Influenza B Mouse Monoclonal Antibody [clone: 8J123]
Anticuerpos
Type: Primary
Antigen: Influenza B
Clonality: monoclonal
Clone: 8J123
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR