Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: US Biological
I7655-65C-FITC
USBII7655-65C-FITCEA 884 EUR
USBII7655-65C-FITC
Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Inositol 1,4,5-trisphosphate 3-kinase A
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR