Anti-Interferon gamma Mouse Monoclonal Antibody [clone: 6A569]

Proveedor: US Biological
I7662-16S
USBII7662-16SEA 565 EUR
USBII7662-16S
Anti-Interferon gamma Mouse Monoclonal Antibody [clone: 6A569]
Anticuerpos
Recommended dilution: Immunohistology: 1:10

Type: Primary
Antigen: Interferon gamma
Clonality: monoclonal
Clone: 6A569
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno IFN gamma
Nombre del antígeno Interferon gamma
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 6A569
Reactividad Human
Huésped Mouse
Isotipo IgG1
Precipitación inmunológica Yes
Forma lyophilized powder
Purification purified by Protein G chromatography
Storage buffer PBS.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR