Anti-Kynurenine 3-monooxygenase Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Proveedor: US Biological
K9800-40-APC
USBIK9800-40-APCEA 884 EUR
USBIK9800-40-APC
Anti-Kynurenine 3-monooxygenase Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: Kynurenine 3-monooxygenase
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR