Anti-Kynurenine 3-monooxygenase Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: US Biological
K9800-40-FITC
USBIK9800-40-FITCEA 884 EUR
USBIK9800-40-FITC
Anti-Kynurenine 3-monooxygenase Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: Kynurenine 3-monooxygenase
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR