Anti-Mouse Monoclonal Antibody [clone: 5E702]

Proveedor: US Biological
L1185-16
USBIL1185-16EA 290 EUR
USBIL1185-16
Anti-Mouse Monoclonal Antibody [clone: 5E702]
Anticuerpos
Recommended dilution: ELISA: 1:50000

Type: Secondary
Antigen: Lambda Light Chain
Clonality: monoclonal
Clone: 5E702
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR