Anti-LSP1 Mouse Monoclonal Antibody

Proveedor: US Biological
L2030-30A-100
USBIL2030-30A-100EA 456 EUR
USBIL2030-30A-100
Anti-LSP1 Mouse Monoclonal Antibody
Anticuerpos
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: LSP1 (lymphocyte-specific protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR