Anti-NADH Dehydrogenase Fe-S Protein 3, 30kD Chicken Polyclonal Antibody

Proveedor: US Biological
N0009-40A
USBIN0009-40AEA 474 EUR
USBIN0009-40A
Anti-NADH Dehydrogenase Fe-S Protein 3, 30kD Chicken Polyclonal Antibody
Anticuerpos
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: NADH Dehydrogenase Fe-S Protein 3, 30kD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR