Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Rhodamine)

Proveedor: US Biological
N2175-91A5
USBIN2175-91A5EA 1160 EUR
USBIN2175-91A5
Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Rhodamine)
Anticuerpos
Type: Primary
Antigen: Neuropeptide B-29
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR