Anti-Nucleoside 5’-diphosphate Kinase Rabbit Polyclonal Antibody

Proveedor: US Biological
N7100-59Y1
USBIN7100-59Y1EA 818 EUR
USBIN7100-59Y1
Anti-Nucleoside 5’-diphosphate Kinase Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Nucleoside 5’-diphosphate Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR