Anti-Nucleoside Diphosphate Kinase 6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: US Biological
N7100-68N-HRP
USBIN7100-68N-HRPEA 884 EUR
USBIN7100-68N-HRP
Anti-Nucleoside Diphosphate Kinase 6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: Nucleoside Diphosphate Kinase 6
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR