Anti-Oxycodone Mouse Monoclonal Antibody [clone: 9L523]

Proveedor: US Biological
O8242-03A
USBIO8242-03AEA 395 EUR
USBIO8242-03A
Anti-Oxycodone Mouse Monoclonal Antibody [clone: 9L523]
Anticuerpos
Type: Primary
Antigen: Oxycodone
Clonality: monoclonal
Clone: 9L523
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR