Anti-PPY Rabbit Polyclonal Antibody

Proveedor: US Biological
P3000-04A
USBIP3000-04AEA 577 EUR
USBIP3000-04A
Anti-PPY Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR