Anti-GOPC Rabbit polyclonal antibody

Proveedor: US Biological
P4207-52
USBIP4207-52EA 647 EUR
USBIP4207-52
Anti-GOPC Rabbit polyclonal antibody
Anticuerpos
Recommended dilution: Western Blot: 1-2?g’ml

Type: Primary
Antigen: GOPC (golgi-associated PDZ and coiled-coil motif containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR