Anti-GOPC Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Proveedor: US Biological
P4207-52B-AP
USBIP4207-52B-APEA 884 EUR
USBIP4207-52B-AP
Anti-GOPC Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: GOPC (golgi-associated PDZ and coiled-coil motif containing)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR