Anti-Platelet Derived Growth Factor BB Rabbit Polyclonal Antibody

Proveedor: US Biological
P4258-17F
USBIP4258-17FEA 822 EUR
USBIP4258-17F
Anti-Platelet Derived Growth Factor BB Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Platelet Derived Growth Factor BB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR