Anti-RAD9A Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: US Biological
R0444-67A-FITC
USBIR0444-67A-FITCEA 884 EUR
USBIR0444-67A-FITC
Anti-RAD9A Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: RAD9A (RAD9 homolog A (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: BH3 domain
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno RAD9A
Nombre del antígeno RAD9 homolog A (S. pombe)
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reactividad Human,Mouse
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
Epítopo BH3 domain
Forma liquid
Purification purified by Protein G chromatography
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Temperatura de almacenamiento Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR