Anti-Resistin-like Molecule, alpha Mouse Monoclonal Antibody [clone: 6D734]

Proveedor: US Biological
R1587-08F
USBIR1587-08FEA 489 EUR
USBIR1587-08F
Anti-Resistin-like Molecule, alpha Mouse Monoclonal Antibody [clone: 6D734]
Anticuerpos
Recommended dilution: ELISA: 1:5000-1:10000); Western Blot: 1:5000-1:10000

Type: Primary
Antigen: Resistin-like Molecule, alpha
Clonality: monoclonal
Clone: 6D734
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR