Anti-RLA, DR Mouse Monoclonal Antibody [clone: 5K114]

Proveedor: US Biological
R2032-50
USBIR2032-50EA 486 EUR
USBIR2032-50
Anti-RLA, DR Mouse Monoclonal Antibody [clone: 5K114]
Anticuerpos
Type: Primary
Antigen: RLA, DR
Clonality: monoclonal
Clone: 5K114
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR