Anti-Src Family, Negative Regulatory Tyrosine Phosphorylation Site Rabbit Polyclonal Antibody

Proveedor: US Biological
S6504-27
USBIS6504-27EA 575 EUR
USBIS6504-27
Anti-Src Family, Negative Regulatory Tyrosine Phosphorylation Site Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Src Family, Negative Regulatory Tyrosine Phosphorylation Site
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR