Anti-STAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: US Biological
S7954-60A-FITC
USBIS7954-60A-FITCEA 884 EUR
USBIS7954-60A-FITC
Anti-STAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:50
Do not freeze!

Type: Primary
Antigen: STAP1 (signal transducing adaptor family member 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR