Anti-THAP11 Goat Polyclonal Antibody

Proveedor: US Biological
T3700-01
USBIT3700-01EA 473 EUR
USBIT3700-01
Anti-THAP11 Goat Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: THAP11 (THAP domain containing 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR