Anti-Vasopressin Receptor, Arg, V1a Rabbit Polyclonal Antibody

Proveedor: US Biological
V2120-32C-100
USBIV2120-32C-100EA 742 EUR
USBIV2120-32C-100
Anti-Vasopressin Receptor, Arg, V1a Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 1-10 ?g/ml

Type: Primary
Antigen: Vasopressin Receptor, Arg, V1a
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR