Anti-ZAP70 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K146]

Proveedor: US Biological
Z0300-10E
USBIZ0300-10EEA 298 EUR
USBIZ0300-10E
Anti-ZAP70 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K146]
Anticuerpos
Type: Primary
Antigen: ZAP70 (zeta-chain (TCR) associated protein kinase 70kDa)
Clonality: monoclonal
Clone: 5K146
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR