uvex silv-Air classic Disposable Particle Filtering Respirators, FFP3
Proveedor: UVEX
8762310 8762312
UVEX8762310EA 78.8 EUR
UVEX8762310 UVEX8762312
uvex silv-Air classic Disposable Particle Filtering Respirators, FFP3
Respiradores Mascarillas desechables Mascarillas para partículas
uvex silv-Air particle filtering preformed mask with exhalation valve with level of protection FFP3.
  • Protection against harmful substances up to 30 times the limit value
  • Soft material edges for better wearer comfort
  • 4-point headband attachment for comfortable, individual and secure fit of the mask
  • Exhalation valve to reduce breathing resistance and to reduce heat and moisture under the mask
  • Integrated, flexible and adjustable nose clip
  • Compatible with uvex safety eyewear
  • All-around sealing lip for high levels of wearer comfort and a reliably secure fit