uvex silv-Air e Particulate Respirator, FPP2/FPP3
Proveedor: UVEX
8787232 8787333 8787317 8787312 8787318 8787330 8787233
UVEX8787232EA 25.7 EUR
UVEX8787232 UVEX8787333 UVEX8787317 UVEX8787312 UVEX8787318 UVEX8787330 UVEX8787233
uvex silv-Air e Particulate Respirator, FPP2/FPP3
Respiradores Mascarillas desechables Mascarillas para partículas
All masks of the uvex silv-Air e family combine our unique technologies of optimum wearer comfort with extremely low breathing resistance. The products are therefore suitable for heavy work and for being worn for long periods of time. The increased size of the filter surface even enables the high-performance models to be worn for several days.
  • High-performance climazone system with three compartments for a comfortable, cool climate inside the mask
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Flexible, adjustable integrated