VARIO® PC 3012 NT VARIO select Chemistry Pumping Unit
Proveedor: VACUUBRAND
25743874 25743851 25743850
VACU25743874EA 13630 EUR
VACU25743874 VACU25743851 VACU25743850
VARIO® PC 3012 NT VARIO select Chemistry Pumping Unit
Bombas Bombas de diafragma
Chemistry diaphragm pumps from VACUUBRAND for aggressive gases and vapors are available in the basic version, with two-point or VARIO® control. Their construction with fluoropolymers makes them very resistant to chemical vapors from inlet to exhaust.
  • VACUU·SELECT vacuum controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times
  • Meets high vacuum requirements for large quantities of most high boiling solvents
  • Extraordinary