Vacuum manifold, Vac Elut™ 20 with tall glass basin
Proveedor: VARIAN
12234104
VARI12234104EA 2560 EUR
VARI12234104
Vacuum manifold, Vac Elut™ 20 with tall glass basin
Colectores
The Vac Elut™ 20 with a large glass basin and collection rack accommodates larger 16×150 mm test tubes. The same high quality material and features on the standard Vac Elut™ system are incorporated on this special unit. These collection vessels can be utilised in combinatorial chemistry applications using large boiling tubes for collection of purified synthesis mixtures, or for any SPE extraction in which an elution volume greater than 10 ml is required.
  • For extractions greater than 10 ml
  • Transparent glass base allows you to monitor the whole collection