HPLC columns, MonoChrom® C18
Proveedor: VARIAN
A0504250X046 A0504300X039 A0504300X040 A0505050X020 A0505100X020 A0505150X046 A0505250X046 A0505MG A0505MG2 A0400030X020 A0400MG2 A0400100X020 A0400150X030 A0400150X046
VARIA0504250X046EA 0 EUR
VARIA0504250X046 VARIA0504300X039 VARIA0504300X040 VARIA0505050X020 VARIA0505100X020 VARIA0505150X046 VARIA0505250X046 VARIA0505MG VARIA0505MG2 VARIA0400030X020 VARIA0400MG2 VARIA0400100X020 VARIA0400150X030 VARIA0400150X046
HPLC columns, MonoChrom® C18
Columnas de cromatografía
MonoChrom® columns are ideal for separation of organic amines, organic acids, amphoteric compounds and neutral compounds. Advanced silica production and silane bonding techniques allow for the production of material with exceptional base-deactivated characteristics.