Filter plates, 96-well, Captiva
Proveedor: VARIAN
A59640002V A596002000 A5960045 A596401000 A5967045 A596002000B A5960045B
VARIA59640002VEA 2060 EUR
VARIA59640002V VARIA596002000 VARIA5960045 VARIA596401000 VARIA5967045 VARIA596002000B VARIA5960045B
Filter plates, 96-well, Captiva
Microplacas Placas de filtro
Filtration is simple, versatile, and necessary to prevent clogging of valuable HPLC columns. Captiva 0,2 μm and 0,45 μm depth filter plates are ideal for filtering samples prior to LC/MS injection. Captiva 10 μm glass fibre filter plates are designed for clarifying highly particle-laden samples, such as freshly thawed plasma and hepatocyte filtration, preventing sample transfer problems from pipette tip clogging. They are perfect for automated systems and for use with DuoSeal 96 96-well seals.
  • Protect HPLC columns from clogging to reduce instrument downtime
  • Clean and clear filtrates